trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98%
|
|
BLDPBD01431594-5GEA
11
EUR
BLDPBD01431594-5G
BLDPBD01431594-100
BLDPBD01431594-25G
trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98%
trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl
Storage condition: Sealed in dry, Room Temperature
Formel:
C₂₀H₂₇F₃ Molecular Weight: 324,43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12911825 CAS-Nummer: 139215-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...