tert-Butyl Hydrogen tetradecanedioate 98+%

BLDPBD01424725-5GEA 285 EUR
BLDPBD01424725-5G BLDPBD01424725-1G BLDPBD01424725-100 BLDPBD01424725-25G
tert-Butyl Hydrogen tetradecanedioate 98+%
tert-Butyl Hydrogen tetradecanedioate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₈H₃₄O₄
Molecular Weight: 314,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23136040
CAS-Nummer: 234082-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR