2-Isobutyl-3-methoxypyrazin 99+%

BLDPBD01414314-100EA 507 EUR
BLDPBD01414314-100 BLDPBD01414314-1G BLDPBD01414314-25G BLDPBD01414314-5G
2-Isobutyl-3-methoxypyrazin 99+%
2-Isobutyl-3-methoxypyrazin

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₄N₂O
Molecular Weight: 166,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006128
CAS-Nummer: 24683-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR