4-(Trifluormethyl)nicotinsäure 98+%
|
|
BLDPBD01412234-100EA
383
EUR
BLDPBD01412234-100
BLDPBD01412234-5G
BLDPBD01412234-25G
BLDPBD01412234-500
4-(Trifluormethyl)nicotinsäure 98+%
4-(Trifluormethyl)nicotinsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082626 CAS-Nummer: 158063-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...