Ospemifene 99% (durch HPLC)
|
|
BLDPBD01407489-250EA
22
EUR
BLDPBD01407489-250
BLDPBD01407489-5G
BLDPBD01407489-1G
Ospemifene 99% (durch HPLC)
Ospemifene
Storage condition: Sealed in dry, 2-8°C
Formel:
C₂₄H₂₃ClO₂ Molecular Weight: 378,90 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00871890 CAS-Nummer: 128607-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...