α,2-Dibrom-5-fluortoluol 98+%
|
|
BLDPBD01405557-25GEA
30
EUR
BLDPBD01405557-25G
BLDPBD01405557-5G
α,2-Dibrom-5-fluortoluol 98+%
α,2-Dibrom-5-fluortoluol
Storage condition: Sealed in dry, 2-8°C
Formel:
C₇H₅Br₂F Molecular Weight: 267,92 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00042187 CAS-Nummer: 112399-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...