4-Mercaptobenzoesäure 90+% (durch HPLC)

BLDPBD01393244-5GEA 205 EUR
BLDPBD01393244-5G BLDPBD01393244-1G BLDPBD01393244-25G
4-Mercaptobenzoesäure 90+% (durch HPLC)
4-Mercaptobenzoic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆O₂S
Molecular Weight: 154,1863 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00016617
CAS-Nummer: 1074-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR