Dithizon AR

Lieferant: BLD PHARMATECH GMBH

Synonyme: 1,5-Diphenylthiocarbazon

BLDPBD01375162-25GEA 43 EUR
BLDPBD01375162-25G BLDPBD01375162-5G
Dithizon AR
Dithizon

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₅NHNHCSN=NC₆H₅
Molecular Weight: 256,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003025
CAS-Nummer: 60-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR