4-(Carboxymethyl)phenylboronsäurepinakolester 98+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD01370860-1GEA 76 EUR
BLDPBD01370860-1G BLDPBD01370860-25G BLDPBD01370860-5G BLDPBD01370860-100
4-(Carboxymethyl)phenylboronsäurepinakolester 98+%
4-(Carboxymethyl)phenylboronsäurepinakolester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₉BO₄
Molecular Weight: 262,1093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038755
CAS-Nummer: 797755-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR