3,4,6-Trichlorpyridazin 90+%

BLDPBD01298777-5GEA 7 EUR
BLDPBD01298777-5G
3,4,6-Trichlorpyridazin 90+%
3,4,6-Trichloropyridazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₄HCl₃N₂
Molecular Weight: 183,4231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00834957
CAS-Nummer: 6082-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR