Tributyl(ethyl)phosphonium Diethyl Phosphate 97%

BLDPBD01120444-100EA 315 EUR
BLDPBD01120444-100 BLDPBD01120444-25G BLDPBD01120444-5G
Tributyl(ethyl)phosphonium Diethyl Phosphate 97%
Tributyl(ethyl)phosphonium Diethyl Phosphate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₈H₄₂O₄P₂
Molecular Weight: 384,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17018975
CAS-Nummer: 20445-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR