2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione 97%
|
|
BLDPBD01104981-250EA
16
EUR
BLDPBD01104981-250
BLDPBD01104981-1G
BLDPBD01104981-5G
BLDPBD01104981-25G
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione 97%
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₃H₂₀O₃ Molecular Weight: 224,30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366927 CAS-Nummer: 172611-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...