2-Bromo-5'-phenyl-1,1':3',1''-terphenyl 98%

BLDPBD01100331-250EA 88 EUR
BLDPBD01100331-250 BLDPBD01100331-1G BLDPBD01100331-100
2-Bromo-5'-phenyl-1,1':3',1''-terphenyl 98%
2-Bromo-5'-phenyl-1,1':3',1''-terphenyl

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₄H₁₇Br
Molecular Weight: 385,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32062871
CAS-Nummer: 1890136-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR