3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole 97%

BLDPBD00964551-1GEA 24 EUR
BLDPBD00964551-1G BLDPBD00964551-250 BLDPBD00964551-5G
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole 97%
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₂H₁₃Br₂N
Molecular Weight: 451,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29072769
CAS-Nummer: 1221237-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR