IR 783 98%

BLDPBD00957967-250EA 48 EUR
BLDPBD00957967-250 BLDPBD00957967-100 BLDPBD00957967-1G BLDPBD00957967-5G
IR 783 98%
IR 783

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₈H₄₆ClN₂NaO₆S₂
Molecular Weight: 749,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093208
CAS-Nummer: 115970-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR