1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulphonyl)imide 95%

BLDPBD00865517-250EA 31 EUR
BLDPBD00865517-250 BLDPBD00865517-25G BLDPBD00865517-1G BLDPBD00865517-5G
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulphonyl)imide 95%
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulphonyl)imide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₈F₂N₂O₅S₂
Molecular Weight: 324,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30749293
CAS-Nummer: 1235234-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR