1-Butylpyridinium bis(trifluoromethanesulphonyl)imide 98%
|
|
BLDPBD00819717-5GEA
23
EUR
BLDPBD00819717-5G
BLDPBD00819717-1G
BLDPBD00819717-25G
BLDPBD00819717-100
1-Butylpyridinium bis(trifluoromethanesulphonyl)imide 98%
1-Butylpyridinium bis(trifluoromethanesulphonyl)imide
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₁H₁₄F₆N₂O₄S₂ Molecular Weight: 416,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16038143 CAS-Nummer: 187863-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...