6-Bromo-8-methoxyisoquinoline 98%

BLDPBD00796725-100EA 158 EUR
BLDPBD00796725-100 BLDPBD00796725-250
6-Bromo-8-methoxyisoquinoline 98%
6-Bromo-8-methoxyisoquinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₈BrNO
Molecular Weight: 238,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124579
CAS-Nummer: 1415564-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR