N-[4-(tert-Butyl)phenyl]-9,9-dimethyl-9H-fluoren-2-amine 98%

BLDPBD00795487-250EA 32 EUR
BLDPBD00795487-250 BLDPBD00795487-1G BLDPBD00795487-5G
N-[4-(tert-Butyl)phenyl]-9,9-dimethyl-9H-fluoren-2-amine 98%
N-[4-(tert-Butyl)phenyl]-9,9-dimethyl-9H-fluoren-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₅H₂₇N
Molecular Weight: 341,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32068070
CAS-Nummer: 944418-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR