(Fmoc-amino)-PEG12-C2-Carboxylic acid 98%

BLDPBD00794385-250EA 68 EUR
BLDPBD00794385-250 BLDPBD00794385-10G BLDPBD00794385-5G
(Fmoc-amino)-PEG12-C2-Carboxylic acid 98%
(Fmoc-amino)-PEG12-C2-Carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₄₂H₆₅NO₁₆
Molecular Weight: 839,97 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08064302
CAS-Nummer: 1952360-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR