1-(2-Propynyloxy)naphthalene 95% (durch GC)

BLDPBD00793326-1GEA 86 EUR
BLDPBD00793326-1G BLDPBD00793326-5G BLDPBD00793326-100 BLDPBD00793326-250
1-(2-Propynyloxy)naphthalene 95% (durch GC)
1-(2-Propynyloxy)naphthalene

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₃H₁₀O
Molecular Weight: 182,22 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03787730
CAS-Nummer: 18542-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR