7-Bromo-2,1,3-benzothiadiazole-4-carbonitrile 98%

BLDPBD00785722-5GEA 1239 EUR
BLDPBD00785722-5G BLDPBD00785722-250 BLDPBD00785722-1G BLDPBD00785722-100
7-Bromo-2,1,3-benzothiadiazole-4-carbonitrile 98%
7-Bromo-2,1,3-benzothiadiazole-4-carbonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₂BrN₃S
Molecular Weight: 240,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28966927
CAS-Nummer: 1331742-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR