9-Benzyl-2-bromo-9H-carbazole 98%

BLDPBD00778764-1GEA 88 EUR
BLDPBD00778764-1G
9-Benzyl-2-bromo-9H-carbazole 98%
9-Benzyl-2-bromo-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₁₄BrN
Molecular Weight: 336,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30469064
CAS-Nummer: 1401863-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR