2-Hexyldecan-1-amine 98% (durch GC, titrimetrische Analyse)

BLDPBD00772525-1GEA 29 EUR
BLDPBD00772525-1G BLDPBD00772525-5G BLDPBD00772525-250 BLDPBD00772525-100
2-Hexyldecan-1-amine 98% (durch GC, titrimetrische Analyse)
2-Hexyldecan-1-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₆H₃₅N
Molecular Weight: 241,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30475384
CAS-Nummer: 62281-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR