1-Butyl-1-methylpiperidinium bis(trifluoromethanesulphonyl)imide 99%
|
|
BLDPBD00772013-100EA
629
EUR
BLDPBD00772013-100
BLDPBD00772013-1G
BLDPBD00772013-25G
BLDPBD00772013-5G
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulphonyl)imide 99%
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulphonyl)imide
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₂H₂₂F₆N₂O₄S₂ Molecular Weight: 436,43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14156017 CAS-Nummer: 623580-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...