Advanced hygienic hand rub, PURELL®
Lieferant: Gojo
PURELL®
GOJO9668-12-EEU00EA 186 EUR
GOJO9668-12-EEU00
Advanced hygienic hand rub, PURELL®
Hautpflege Seifen und Hautreiniger
No-rinse hygienic hand rub feels great to use and is clinically proven to maintain skin health.
  • Includes moisturisers to help keep skin healthy
  • Kills 99,99% of most common germs that may be harmful
  • Alcohol formulation 70% is suitable for frequent use by hospital staff
  • Food taint test compliant: BS EN ISO 4120:2007
Recommended for use on hands that are lightly soiled, and in conjunction with a hand hygiene regime.
Bactericidal according to EN 1500 (hand sanitiser). Conforms to EN 12791 (surgical rub), EN 1276 and EN 13727, virucidal according to EN 14476, fungicidal and yeasticidal according to EN 13624 and EN 1650; and mycobactericidal according to EN 14348.