BSOCOES (Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulphone)
Lieferant: G Bioscienes
BC01EA 448 EUR
BC01
BSOCOES (Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulphone)
BSOCOES (Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulphone)
(Bis(2-[Succinimidooxycarbonyloxy]ethyl) sulfone Homobifunctional Cross Linkers.
Formel: C₁₄H₁₆N₂O₁₂S
Molecular Weight: 436,35 g/mol
MDL: MFCD00047123
CAS-Nummer: 57683-72-4
Bestellen
Spezifikation Testergebnisse
Spacer Arm (Å) 13
Reactive Toward Primary Amines
Membrane Permeable YES
Water Soluble NO
Cleavable/ Reversible Base