qPCR and go probe No-ROX kit
Lieferant: MP Biomedicals
11EBI06200EA 1294 EUR
11EBI06200 11EBI06050
qPCR and go probe No-ROX kit
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
Fast and sensitive qPCR Probe Master Mix. For all other platforms, example: Roche, Bio-Rad, Qiagen and more.
  • Reproducible, robust, sensitive, fast, efficient, specific, excellent performance in multiplex assays
  • Store at –20 °C
The qPCRand Go Probe No-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments. The kit has been formulated for use with probe-detection technology, including TaqMan®, Scorpions® and molecular beacon probes. A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCRand Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR. qPCRand Go Probe is provided as a 2X mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Standards: ISO13485 certified.