Peptide cleanup pipette tips
Lieferant: Agilent Technologies
AGLS5188-5239EA 214 EUR
AGLS5188-5239
Peptide cleanup pipette tips
Peptide cleanup pipette tips are packed with ZORBAX SB-C18 silica used for purification and concentration of peptide, protein digests, proteins, or oligonucleotide samples by pipetting sample through the tip. Typical applications include sample preparation for analysis using ESI and MALDI-MS.
  • C18-silica solid phase extraction material is embedded into the wall of the pipette tip (no glue or polymer matrix is used)
  • Maximizing surface area in contact with the sample and providing excellent flow properties
  • Uses an easy-to-follow protocol that provides highly reproducible recoveries of peptides from in-gel digests with low interferences for mass spectrometry (MS) analysis
Recommended one-time use only. Designed to fit all standard 10 µl adjustable pipettes.
Bitte beachten Research use only.