SaniSure® Single-Use Sanitary Clamps, Nylon
Lieferant: SaniSure
MFLX30562-22EA 34.7 EUR
MFLX30562-22 MFLX30562-21
SaniSure® Single-Use Sanitary Clamps, Nylon
Klemmen Schlauchklemmen und Sanitärklemmen
Fast, safe, secure, and cost-effective connections!
  • Even 360-degree compression to prevent leakage
  • Validatable closure torque
  • Withstands pressures up to 100 psi
  • Reduces installation time
The SaniSure® clamps feature an innovative design for fast, safe, secure connections and offer a cost-effective solution for connecting single-use installations. Made of USP Class VI-approved nylon, these clamps are designed for single use applications but are durable enough to withstand repetitive use.
Unlike traditional two-point load stainless steel sanitary clamps, the design of these clamps ensure even 360-degree compression over the entire fitting surface—no more leaks and no gasket intrusion in your process line! Their low-profile rounded edges reduce the risk of penetrating bag assemblies, and the evenly controlled compression allows easy connection for plastic-to-plastic, plastic-to-stainless steel, or stainless steel-to-stainless steel coupling joints.
To use, simply insert the cable tie end until its made a couple clicks; position the clamp over the ferrule; hold both clamp parts together; move the clamp around the tri-clamp fitting joint and slide the clamp closed. Complete the closure by using the clamping tool (not included) to bring it to the proper tension.
The easy-to-use design reduces the chances of risk associated with carpal tunnel, or other wrist injuries, and yields a tamper-proof connection that is not operator-dependent. As a single-use device, they eliminate the need for cleaning validations and reduce your overall downtime for installation.