Finish Thompson Economical Drum Pump Tube Sets
Lieferant: Finish Thompson
MFLX72004-67EA 487 EUR
MFLX72004-67 MFLX72004-69 MFLX72004-63 MFLX72004-64
Finish Thompson Economical Drum Pump Tube Sets
Pumpen Fass/Transferpumpen
Customize your drum pump with a wide range of tube options.
  • Choose between polypropylene (PP) and stainless steel (SS) tube set options
Polypropylene tube sets are ideal for economical general lab usage. Pure polypropylene/PVDF tube sets are suitable for chemical resistant applications, and stainless steel tube sets are durable for industrial use.