Distek, Inc. has revolutionised bench top scale bioreactors with the release of our BIOne 1250 bioprocess control station with autoclavable bioreactor for mammalian models. The sleek appearance and intuitive touch screen interface are only the beginning. The BIOne 1250 is designed to be the easiest to use and most capable stand-alone controller on the market.
- Touch screen interface
- Smart media filling
- Email and text alerts
- Trend up to eight parameters
- Variable speed pumps
- 2 and 5 L working volume
- Configurable for your lab
Control all bioreactor parameters directly from the BIOne 1250 interface, or, for remote control, use our optional OPC server and communicate with your choice of SCADA system.
From the pop-up tab of your selected parameter, control set points, calibrations, alarms and more. Reduce development time lines by utilising the live, dual-graph trend screen. Analyse and compare current and previous runs. Track all optimised changes and export data to further analyse.
Standards: ETL, CE
Bestellinformation: Includes touch screen display, vessel, head plate, stand, heater jacket, four pumps, pH and DO sensors plus accessory kit. Includes a one year comprehensive warranty. Does not include installation or qualification services.