RNase A + RNase (from Aspergillus oryzae), RNace-It
Lieferant: Agilent Technologies
AGLS400720EA 247 EUR
AGLS400720
RNase A + RNase (from Aspergillus oryzae), RNace-It
Enzyme
Purified Ribonuclease Cocktail tested to be free of contaminating activity for the degradation of nucleic acids.
  • Contains: RNase A (2 mg/ml), nonspecific RNase from Aspergillus oryzae (4 units/µl)
  • Unit Definition: 20 units will degrade the RNA in a 1,5 ml plasmid preparation
  • Reaction Conditions: Active in any neutral buffer (pH 6,0 to 9,0) or water with sodium chloride concentrations ˂100 mM
RNace-IT ribonuclease cocktail is a mixture of two RNases: RNase A and a nonspecific RNase from Aspergillus oryzae known as RNase T1. The combined action of these ribonucleases results in the degradation of RNA into small oligoribonucleotides.
Bitte beachten For research use only. Not for use in diagnostic procedures.