Affinity removal spin cartridges HSA
Lieferant: Agilent Technologies
AGLS5188-5334EA 500 EUR
AGLS5188-5334 HEWL5188-5334
Affinity removal spin cartridges HSA
Protein-Aufbereitungs Protein Purification Kits
The affinity removal spin cartridge – human albumin is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Compatible with standard benchtop microcentrifuges and Luer-lok syringes
  • Serum load volume is 65 µl per run
  • Optimise sample recovery by utilising our rapid, single injection protocols and optimised buffers
Removal of albumin improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.