High-recovery protein columns, mRP-C18
Lieferant: Agilent Technologies
HEWL5188-6510EA 0 EUR
HEWL5188-6510
High-recovery protein columns, mRP-C18
Chromatographiesäulen
The mRP-C18 high-recovery protein column is an LC column designed for sample desalting, followed by chromatographic separation or fractionation of the complex protein samples (like immunodepleted serum or plasma proteins) with extremely high recovery of sample when compared to conventional RP HPLC columns.
  • Greater than 95 to 99% protein sample recovery has been observed with immunodepleted serum
  • Can load up to 380 µg of total protein mass without reducing chromatographic resolution of the proteins
  • Column packed with macroporous C18-bonded ultrapure 5 µm particle silica designed to reduce or eliminate strong adsorption of proteins
  • Maximum operating pressure of 4000 psi (250 bar)
Compatible with water and all common organic solvents.