Antibodies, Anti-FLAG®
Lieferant: Agilent Technologies
AGLS200474EA 411 EUR
AGLS200474 AGLS200473
Antibodies, Anti-FLAG®
Antikörper
The FLAG® peptide (DYKDDDDK) is widely used as an epitope tag for protein expression analysis, purification, and detection.
  • Rat monoclonal antibody to DYKDDDDK epitope tag
  • High specificity – reduced background and less cross-reactivity
  • Highly sensitivity – strong signal and ability to detect low amounts of tag
The Anti-FLAG® M2 Antibody is optimized for the detection of FLAG-fusion proteins in mammalian, plant and bacterial expression systems. This antibody provides equivalent sensitivity and specificity to similar antibodies.
Bitte beachten For research use only. Not for use in diagnostic procedures.