4-Amino-3,5-difluorphenol

Lieferant: Apollo Scientific
Achtung

APOSPC8306-1GEA 364 EUR
APOSPC8306-1G
4-Amino-3,5-difluorphenol
4-Amino-3,5-difluorphenol
Formel: C₆H₅F₂NO
Molecular Weight: 145,11 g/mol
Schmelzpunkt: 144…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094491
CAS-Nummer: 135086-76-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR