N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
Lieferant: Apollo Scientific
APOSPC8081-1GEA
212
EUR
APOSPC8081-1G
N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
Formel:
C₁₄H₁₁BrF₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800987 CAS-Nummer: 951885-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...