2-Brom-N-cyclopentyl-5-fluorbenzamid

Lieferant: Apollo Scientific

APOSPC8018-1GEA 176 EUR
APOSPC8018-1G
2-Brom-N-cyclopentyl-5-fluorbenzamid
2-Brom-N-cyclopentyl-5-fluorbenzamid
Formel: C₁₀H₉BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09800910
CAS-Nummer: 951884-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR