2-Brom-N-butyl-5-fluorbenzamid

Lieferant: Apollo Scientific

APOSPC7976-1GEA 151 EUR
APOSPC7976-1G
2-Brom-N-butyl-5-fluorbenzamid
2-Brom-N-butyl-5-fluorbenzamid
Formel: C₁₁H₁₃BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09800917
CAS-Nummer: 951884-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR