3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC7183-1GEA
156
EUR
APOSPC7183-1G
3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure 98%
3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₁BFNO₄ Molecular Weight: 227,00 Schmelzpunkt: 217…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689535 CAS-Nummer: 874289-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...