Tetrapropylammoniumtetrafluoroborat 98%

Lieferant: Apollo Scientific
Achtung

APOSPC6941-25GEA 280 EUR
APOSPC6941-25G
Tetrapropylammoniumtetrafluoroborat 98%
Tetrapropylammonium tetrafluoroborate
Formel: C₁₂H₂₈BF₄N
Molecular Weight: 273,16 g/mol
Schmelzpunkt: 245…247 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 338-38-5
EINECS: 206-416-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR