N-Methyl-4-fluor-2-nitroanilin

Lieferant: Apollo Scientific

Achtung

APOSPC6723-1GEA 19 EUR
APOSPC6723-1G
N-Methyl-4-fluor-2-nitroanilin
N-Methyl-4-fluor-2-nitroanilin
Formel: C₇H₇FN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD00792440
CAS-Nummer: 704-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR