3-Iod-4-(trifluormethoxy)benzoesäure 95+%

Lieferant: Apollo Scientific

Achtung

APOSPC502616-250MGEA 127 EUR
APOSPC502616-250MG
3-Iod-4-(trifluormethoxy)benzoesäure 95+%
3-Iodo-4-(trifluoromethoxy)benzoic acid
Formel: C₈H₄F₃IO₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1110709-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR