4-Fluor-(3-methoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

APOSPC5015-1GEA 32.1 EUR
APOSPC5015-1G
4-Fluor-(3-methoxycarbonyl)phenylboronsäure 98%
4-Fluor-(3-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197,96 g/mol
Schmelzpunkt: 253…255 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06801688
CAS-Nummer: 874219-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR