3-(Dimethylcarbamoyl)-4-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

APOSPC4998-1GEA 299 EUR
APOSPC4998-1G
3-(Dimethylcarbamoyl)-4-fluorphenylboronsäure 98%
3-(Dimethylcarbamoyl)-4-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211,00 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08235046
CAS-Nummer: 874219-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR