2-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 95+%
Lieferant: Apollo Scientific
APOSPC412493-1GEA
77.1
EUR
APOSPC412493-1G
2-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 95+%
2-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester
Formel:
C₁₃H₁₅BF₄O₂ Molecular Weight: 290,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458187 CAS-Nummer: 1073353-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...