3-Fluor-4-(trifluormethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
APOSPC412492-1GEA
91.6
EUR
APOSPC412492-1G
3-Fluor-4-(trifluormethyl)phenylboronsäure 98%
3-Fluor-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207,92 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08706261 CAS-Nummer: 864759-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...