2,5-Difluor-p-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC400739-5GEA
101
EUR
APOSPC400739-5G
2,5-Difluor-p-toluylsäure
2,5-Difluor-p-toluylsäure
Formel:
C₈H₆F₂O₂ Molecular Weight: 172,13 g/mol Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08063230 CAS-Nummer: 103877-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...