Kalium(3-phenylpropyl)trifluoroborat

Lieferant: Apollo Scientific
Achtung

APOSPC400014-1GEA 85.2 EUR
APOSPC400014-1G
Kalium(3-phenylpropyl)trifluoroborat
Potassium-3-phenylpropyltrifluoroborate
Formel: C₉H₁₁BF₃K
Lagertemperatur: Raumtemperatur
MDL: MFCD09993266
CAS-Nummer: 329976-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR